4-Fluoro-3-(5-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,4-oxadiazol-3-yl)benzaldehyde

Lieferant: Apollo Scientific

PC430588-250MG
APOSPC430588-250MGEA 587 EUR
APOSPC430588-250MG
4-Fluoro-3-(5-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,4-oxadiazol-3-yl)benzaldehyde
4-Fluoro-3-(5-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,4-oxadiazol-3-yl)benzaldehyde
Formel: C₁₄H₁₁FN₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18157701
CAS-Nummer: 1258652-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR