2,3-Diamino-n-(4-fluoro-3-(trifluoromethyl)phenyl)benzamide

Lieferant: Apollo Scientific

PC430589-250MG
APOSPC430589-250MGEA 587 EUR
APOSPC430589-250MG
2,3-Diamino-n-(4-fluoro-3-(trifluoromethyl)phenyl)benzamide
2,3-Diamino-n-(4-fluoro-3-(trifluoromethyl)phenyl)benzamide
Formel: C₁₄H₁₁F₄N₃O
Molecular Weight: 313,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23378558
CAS-Nummer: 1421933-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR