2,3-Diamino-N-(4-(trifluoromethyl)phenyl)benzamide

Lieferant: Apollo Scientific

PC430590-250MG
APOSPC430590-250MGEA 587 EUR
APOSPC430590-250MG
2,3-Diamino-N-(4-(trifluoromethyl)phenyl)benzamide
2,3-Diamino-N-(4-(trifluoromethyl)phenyl)benzamide
Formel: C₁₄H₁₂F₃N₃O
Molecular Weight: 295,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23378557
CAS-Nummer: 1421933-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR