8-Fluoroquinazolin-2-amine

Lieferant: Apollo Scientific

PC430591-500MG
APOSPC430591-500MGEA 587 EUR
APOSPC430591-500MG
8-Fluoroquinazolin-2-amine
8-Fluoroquinazolin-2-amine
Formel: C₈H₆FN₃
Molecular Weight: 163,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193275
CAS-Nummer: 190274-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR