N-Succinimidyl trifluoroacetate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: TFA-NHS, N-(Trifluoroacetoxy)succinimide, Trifluoroacetic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 2,2,2-trifluoroacetate

PC430592-1G
APOSPC430592-1GEA 101 EUR
APOSPC430592-1G
N-Succinimidyl trifluoroacetate 95%
N-Succinimidyl trifluoroacetate
Formel: C₆H₄F₃NO₄
Molecular Weight: 211,10 g/mol
Schmelzpunkt: 176 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00595252
CAS-Nummer: 5672-89-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR