3-(6-Fluoropyridin-3-yl)-5-methylbenzaldehyde

Lieferant: Apollo Scientific

PC430593-500MG
APOSPC430593-500MGEA 587 EUR
APOSPC430593-500MG
3-(6-Fluoropyridin-3-yl)-5-methylbenzaldehyde
3-(6-Fluoropyridin-3-yl)-5-methylbenzaldehyde
Formel: C₁₃H₁₀FNO
Molecular Weight: 215,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585334
CAS-Nummer: 1206969-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR