(2-Fluoro-phenyl)-(octahydro-[1,6]naphthyridin-1-yl)-methanone

Lieferant: Apollo Scientific

PC430594-500MG
APOSPC430594-500MGEA 587 EUR
APOSPC430594-500MG
(2-Fluoro-phenyl)-(octahydro-[1,6]naphthyridin-1-yl)-methanone
(2-Fluoro-phenyl)-(octahydro-[1,6]naphthyridin-1-yl)-methanone
Formel: C₁₅H₁₉FN₂O
Molecular Weight: 262,323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585328
CAS-Nummer: 1820572-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR