3-(3,5-Dichlorophenylcarbamoyl)-2-fluorobenzoic acid

Lieferant: Apollo Scientific

PC430595-500MG
APOSPC430595-500MGEA 587 EUR
APOSPC430595-500MG
3-(3,5-Dichlorophenylcarbamoyl)-2-fluorobenzoic acid
3-(3,5-Dichlorophenylcarbamoyl)-2-fluorobenzoic acid
Formel: C₁₄H₈Cl₂FNO₃
Molecular Weight: 328,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585330
CAS-Nummer: 1206969-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR