2-(2,2,2-Trifluoroethoxy)-6-chloropyridine

Lieferant: Apollo Scientific

PC430596-500MG
APOSPC430596-500MGEA 598 EUR
APOSPC430596-500MG
2-(2,2,2-Trifluoroethoxy)-6-chloropyridine
2-(2,2,2-Trifluoroethoxy)-6-chloropyridine
Formel: C₇H₅ClF₃NO
Molecular Weight: 211,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077489
CAS-Nummer: 77145-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR