3-(3-Fluorophenyl)-1,2,4-thiadiazol-5(4H)-one

Lieferant: Apollo Scientific

PC430598-500MG
APOSPC430598-500MGEA 671 EUR
APOSPC430598-500MG
3-(3-Fluorophenyl)-1,2,4-thiadiazol-5(4H)-one
3-(3-Fluorophenyl)-1,2,4-thiadiazol-5(4H)-one
Formel: C₈H₅FN₂OS
Molecular Weight: 196,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585315
CAS-Nummer: 1206969-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR