Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific

PC430599-500MG
APOSPC430599-500MGEA 671 EUR
APOSPC430599-500MG
Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate
Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate
Formel: C₂₀H₁₈F₃N₃O₄S
Molecular Weight: 453,435 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585471
CAS-Nummer: 1206970-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR