5-(2,2,2-Trifluoroacetamido)-2-methoxy-4-nitrophenyl pivalate

Lieferant: Apollo Scientific

PC430600-500MG
APOSPC430600-500MGEA 757 EUR
APOSPC430600-500MG
5-(2,2,2-Trifluoroacetamido)-2-methoxy-4-nitrophenyl pivalate
5-(2,2,2-Trifluoroacetamido)-2-methoxy-4-nitrophenyl pivalate
Formel: C₁₄H₁₅F₃N₂O₆
Molecular Weight: 364.274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27635168
CAS-Nummer: 660870-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR