2-(3-(Trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)-n-(prop-2-ynyl)acetamide

Lieferant: Apollo Scientific

PC430602-500MG
APOSPC430602-500MGEA 754 EUR
APOSPC430602-500MG
2-(3-(Trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)-n-(prop-2-ynyl)acetamide
2-(3-(Trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)-n-(prop-2-ynyl)acetamide
Formel: C₁₃H₁₄F₃N₃O
Molecular Weight: 285,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157703
CAS-Nummer: 1258652-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR