1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
Lieferant: Apollo Scientific
PC430603-1G
APOSPC430603-1GEA
797
EUR
APOSPC430603-1G
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
Formel:
C₁₄H₂₁FN₂O Molecular Weight: 252,328 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585244 CAS-Nummer: 1206970-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...