1-(4-Bromo-2-fluorobenzyl)piperidine
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-fluorophenyl-1-(piperidinomethyl)benzen
PC430606-1G
APOSPC430606-1GEA
974
EUR
APOSPC430606-1G
1-(4-Bromo-2-fluorobenzyl)piperidine
1-(4-Bromo-2-fluorobenzyl)piperidine
Formel:
C₁₂H₁₅BrFN Molecular Weight: 272,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14645209 CAS-Nummer: 1200131-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...