(4-Bromo-2-fluorophenyl)-n,n-dimethylmethanamine

Lieferant: Apollo Scientific

Synonyme: 1-(4-Bromo-2-fluorophenyl)-N,N-dimethylmethanamine, (4-Bromo-2-fluoro-benzyl)-dimethyl-amine

PC430607-1G
APOSPC430607-1GEA 974 EUR
APOSPC430607-1G
(4-Bromo-2-fluorophenyl)-n,n-dimethylmethanamine
(4-Bromo-2-fluorophenyl)-n,n-dimethylmethanamine
Formel: C₉H₁₁BrFN
Molecular Weight: 232,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795894
CAS-Nummer: 1159976-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR