Ethyl 7-fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylate
Lieferant: Apollo Scientific
PC430609-1G
APOSPC430609-1GEA
1020
EUR
APOSPC430609-1G
Ethyl 7-fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylate
Ethyl 7-fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylate
Formel:
C₁₃H₁₂FNO₄ Molecular Weight: 265,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18157738 CAS-Nummer: 622369-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...