4-Nitro-2-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC4317-5G
APOSPC4317-5GEA 24.8 EUR
APOSPC4317-5G
4-Nitro-2-(trifluormethyl)benzonitril 98%
4-Nitro-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
Molecular Weight: 216,12 g/mol
Schmelzpunkt: 50…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075729
CAS-Nummer: 320-47-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR