2-(Trifluoromethylthio)phenylacetic acid 98%

Lieferant: Apollo Scientific
Achtung

PC4324-250MG
APOSPC4324-250MGEA 109 EUR
APOSPC4324-250MG
2-(Trifluoromethylthio)phenylacetic acid 98%
2-(Trifluoromethylthio)phenylacetic acid
Formel: C₉H₇F₃O₂S
Molecular Weight: 236,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236349
CAS-Nummer: 239080-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR