3-(Trifluoromethyl)phenylurea 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4344-10G
PC4344-1G
PC4344-50G
PC4344-100G
APOSPC4344-10GEA
29
EUR
APOSPC4344-10G
APOSPC4344-1G
APOSPC4344-50G
APOSPC4344-100G
3-(Trifluoromethyl)phenylurea 98%
3-(Trifluoromethyl)phenylurea
Formel:
C₈H₇F₃N₂O Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025424 CAS-Nummer: 13114-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...