3-(Difluoromethoxy)phenol 95%

Lieferant: Apollo Scientific
Gefahr

PC4356-1G
APOSPC4356-1GEA 105 EUR
APOSPC4356-1G
3-(Difluoromethoxy)phenol 95%
3-(Difluoromethoxy)phenol

Not to be stored in glass

Formel: C₇H₆F₂O₂
Molecular Weight: 160,118 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236235
CAS-Nummer: 88798-13-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR