N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 95%

Lieferant: Apollo Scientific

Synonyme: Boc-Phe(3,4-F)-OH, Boc-Phe(3,4-DiF)-OH, N-Boc-3,4-difluoro-L-phenylalanine

PC4360-10G PC4360-1G PC4360-5G
APOSPC4360-10GEA 778 EUR
APOSPC4360-10G APOSPC4360-1G APOSPC4360-5G
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 95%
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine
Formel: C₁₄H₁₇F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00672518
CAS-Nummer: 198474-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR