2,2-Difluor-1,3-benzodioxol-5-carbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid
PC4367-250MG
APOSPC4367-250MGEA
21.9
EUR
APOSPC4367-250MG
2,2-Difluor-1,3-benzodioxol-5-carbonsäure 98%
2,2-Difluor-1,3-benzodioxol-5-carbonsäure
Formel:
C₈H₄F₂O₄ Molecular Weight: 202,12 g/mol Schmelzpunkt: 153…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792417 CAS-Nummer: 656-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...