1-(4-Trifluoromethylphenyl)-1,2-propandione 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC4369-1G
APOSPC4369-1GEA
72.5
EUR
APOSPC4369-1G
1-(4-Trifluoromethylphenyl)-1,2-propandione 95+%
1-(4-Trifluoromethylphenyl)-1,2-propandione
Formel:
C₁₀H₇F₃O₂ Siedepunkt: 70 °C/0,2mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941310 CAS-Nummer: 10557-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...