2'-Fluor-3'-(trifluormethyl)acetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone

PC4371K-1G PC4371K-25G PC4371K-5G
APOSPC4371K-1GEA 21.9 EUR
APOSPC4371K-1G APOSPC4371K-25G APOSPC4371K-5G
2'-Fluor-3'-(trifluormethyl)acetophenon 98%
2'-Fluor-3'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206.14 g/mol
Dichte: 1.02 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00061145
CAS-Nummer: 207853-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR