2'-Fluor-3'-(trifluormethyl)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone
PC4371K-1G
PC4371K-25G
PC4371K-5G
APOSPC4371K-1GEA
21.9
EUR
APOSPC4371K-1G
APOSPC4371K-25G
APOSPC4371K-5G
2'-Fluor-3'-(trifluormethyl)acetophenon 98%
2'-Fluor-3'-(trifluormethyl)acetophenon
Formel:
C₉H₆F₄O Dichte: 1,02 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061145 CAS-Nummer: 207853-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...