2'-Fluor-5'-(trifluormethyl)acetophenon 98%

Lieferant: Apollo Scientific
Achtung

PC4371M-25G PC4371M-5G PC4371M-100G
APOSPC4371M-25GEA 145 EUR
APOSPC4371M-25G APOSPC4371M-5G APOSPC4371M-100G
2'-Fluor-5'-(trifluormethyl)acetophenon 98%
2'-Fluor-5'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061248
CAS-Nummer: 202664-53-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR