3-Fluor-5-(trifluormethyl)benzophenon 97%

Lieferant: Apollo Scientific
Achtung

PC4374J-1G
APOSPC4374J-1GEA 36.4 EUR
APOSPC4374J-1G
3-Fluor-5-(trifluormethyl)benzophenon 97%
3-Fluor-5-(trifluormethyl)benzophenon
Formel: C₁₄H₈F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00061256
CAS-Nummer: 239087-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR