3-Fluor-5-(trifluormethyl)benzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4375I-1G
PC4375I-10G
PC4375I-5G
PC4375I-25G
APOSPC4375I-1GEA
21.9
EUR
APOSPC4375I-1G
APOSPC4375I-10G
APOSPC4375I-5G
APOSPC4375I-25G
3-Fluor-5-(trifluormethyl)benzylalkohol 97%
3-Fluor-5-(trifluormethyl)benzylalkohol
Formel:
C₈H₆F₄O Siedepunkt: 80…82 °C/3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061169 CAS-Nummer: 184970-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...