4-Fluor-2-(trifluormethyl)benzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Fluoro-2-(trifluoromethyl)phenyl)methanol

PC4375J-1G PC4375J-5G PC4375J-25G
APOSPC4375J-1GEA 21.9 EUR
APOSPC4375J-1G APOSPC4375J-5G APOSPC4375J-25G
4-Fluor-2-(trifluormethyl)benzylalkohol 97%
4-Fluor-2-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
Molecular Weight: 194.13 g/mol
Schmelzpunkt: 55…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061170
CAS-Nummer: 220227-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR