4-Fluor-2-(trifluormethyl)benzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-2-(trifluoromethyl)phenyl)methanol
PC4375J-1G
PC4375J-5G
PC4375J-25G
APOSPC4375J-1GEA
21.9
EUR
APOSPC4375J-1G
APOSPC4375J-5G
APOSPC4375J-25G
4-Fluor-2-(trifluormethyl)benzylalkohol 97%
4-Fluor-2-(trifluormethyl)benzylalkohol
Formel:
C₈H₆F₄O Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061170 CAS-Nummer: 220227-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...