2-Fluor-5-(trifluormethyl)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC4376G-1G PC4376G-5G PC4376G-25G
APOSPC4376G-1GEA 0 EUR
APOSPC4376G-1G APOSPC4376G-5G APOSPC4376G-25G
2-Fluor-5-(trifluormethyl)benzylbromid 97%
2-Fluor-5-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00061174
CAS-Nummer: 220239-69-0
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR