2-Fluor-5-(trifluormethyl)benzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC4376G-1G
PC4376G-5G
PC4376G-25G
APOSPC4376G-1GEA
0
EUR
APOSPC4376G-1G
APOSPC4376G-5G
APOSPC4376G-25G
2-Fluor-5-(trifluormethyl)benzylbromid 97%
2-Fluor-5-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061174 CAS-Nummer: 220239-69-0 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...