4-Fluor-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC4376J-10G PC4376J-100G PC4376J-50G
APOSPC4376J-10GEA 21.9 EUR
APOSPC4376J-10G APOSPC4376J-100G APOSPC4376J-50G
4-Fluor-2-(trifluormethyl)benzylbromid 98%
4-Fluor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00061176
CAS-Nummer: 206860-48-2
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR