2-Fluor-4-(trifluormethyl)phenylacetonitril 94%

Lieferant: Apollo Scientific
Gefahr

PC4377F-10G PC4377F-5G PC4377F-1G
APOSPC4377F-10GEA 0 EUR
APOSPC4377F-10G APOSPC4377F-5G APOSPC4377F-1G
2-Fluor-4-(trifluormethyl)phenylacetonitril 94%
2-Fluor-4-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD00061179
CAS-Nummer: 239087-11-7
UN: 3276
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR