2-Fluor-5-(trifluormethyl)phenylacetonitril 98%

Lieferant: Apollo Scientific
Gefahr

PC4377G-10G PC4377G-5G PC4377G-1G
APOSPC4377G-10GEA 0 EUR
APOSPC4377G-10G APOSPC4377G-5G APOSPC4377G-1G
2-Fluor-5-(trifluormethyl)phenylacetonitril 98%
2-Fluor-5-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD00061180
CAS-Nummer: 220227-59-8
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR