3-Fluor-5-(trifluormethyl)phenylacetonitril 98%

Lieferant: Apollo Scientific
Achtung

PC4377J-10G PC4377J-5G PC4377J-1G
APOSPC4377J-10GEA 0 EUR
APOSPC4377J-10G APOSPC4377J-5G APOSPC4377J-1G
3-Fluor-5-(trifluormethyl)phenylacetonitril 98%
3-Fluor-5-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD00061181
CAS-Nummer: 239087-12-8
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR