2'-Fluor-4'-(trifluormethyl)propiophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4377Q-1G
APOSPC4377Q-1GEA
21.9
EUR
APOSPC4377Q-1G
2'-Fluor-4'-(trifluormethyl)propiophenon 97%
2'-Fluor-4'-(trifluormethyl)propiophenon
Formel:
C₁₀H₈F₄O Molecular Weight: 220.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061259 CAS-Nummer: 208173-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...