2'-Fluor-5'-(trifluormethyl)propiophenon 97%

Lieferant: Apollo Scientific
Achtung

PC4377R-1G
APOSPC4377R-1GEA 21.9 EUR
APOSPC4377R-1G
2'-Fluor-5'-(trifluormethyl)propiophenon 97%
2'-Fluor-5'-(trifluormethyl)propiophenon
Formel: C₁₀H₈F₄O
Molecular Weight: 220,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061260
CAS-Nummer: 207974-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR