5-Fluoruracil 99%

Lieferant: Apollo Scientific
Gefahr

PC4380-100G PC4380-25G
APOSPC4380-100GEA 0 EUR
APOSPC4380-100G APOSPC4380-25G
5-Fluoruracil 99%
5-Fluoruracil

Antineoplastic agent. Not USP. Not for drug use.

Formel: C₄H₃FN₂O₂
Siedepunkt: 190…200 °C/0,1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00006018
CAS-Nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR