Fomblin® Y L-VAC 14/6

Lieferant: Apollo Scientific
Achtung

PC4382-1KG
APOSPC4382-1KGEA 621 EUR
APOSPC4382-1KG
Fomblin® Y L-VAC 14/6
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized

A linear perfluoro-polyether fw approx. 2500.

Dichte: 1,9 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00163696
CAS-Nummer: 69991-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR