2-[(4-Ethoxyphenyl)amino]-4-(4-fluoro-3-methylphenyl)-1,3-thiazole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-ethoxyphenyl)-4-(4-fluoro-3-methylphenyl)-1,3-thiazol-2-amine

PC4385-100MG
APOSPC4385-100MGEA 94.4 EUR
APOSPC4385-100MG
2-[(4-Ethoxyphenyl)amino]-4-(4-fluoro-3-methylphenyl)-1,3-thiazole 98%
2-[(4-Ethoxyphenyl)amino]-4-(4-fluoro-3-methylphenyl)-1,3-thiazole
Formel: C₁₈H₁₇FN₂OS
Molecular Weight: 328,404 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00129327
CAS-Nummer: 1092346-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR