Gadolinium(III)fluorid 99,9%

Lieferant: Apollo Scientific

Synonyme: Gadoliniumtrifluorid

PC4386P-100G PC4386P-50G PC4386P-10G
APOSPC4386P-100GEA 335 EUR
APOSPC4386P-100G APOSPC4386P-50G APOSPC4386P-10G
Gadolinium(III)fluorid 99,9%
Gadolinium(III)fluorid
Formel: F₃Gd
Lagertemperatur: Raumtemperatur
MDL: MFCD00016113
CAS-Nummer: 13765-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR