(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol

Lieferant: Apollo Scientific

Synonyme: (R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-[1,1'-binaphthalene]-2,2'-diol, (R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]1,1'-binaphthalene-2,2'-diol

PC440000-250MG
APOSPC440000-250MGEA 154 EUR
APOSPC440000-250MG
(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
Formel: C₃₆H₁₈F₁₂O₂
Molecular Weight: 710,508 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689862
CAS-Nummer: 756491-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR