(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol

Lieferant: Apollo Scientific

PC440001-250MG
APOSPC440001-250MGEA 0 EUR
APOSPC440001-250MG
(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
Formel: C₃₆H₁₈F₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08689862
CAS-Nummer: 849939-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR