(S)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
Lieferant: Apollo Scientific
|
Achtung
|
PC440003-250MG
APOSPC440003-250MGEA
582
EUR
APOSPC440003-250MG
(S)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
(S)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
Formel:
C₃₂H₁₈F₄O₂ Molecular Weight: 510,478 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17019237 CAS-Nummer: 1706439-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...