(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate
PC440004-250MG
APOSPC440004-250MGEA
169
EUR
APOSPC440004-250MG
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel:
C₃₆H₁₇F₁₂O₄P Molecular Weight: 772,472 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689863 CAS-Nummer: 791616-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...