(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific
Achtung

Synonyme: (11bS)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

PC440005-250MG
APOSPC440005-250MGEA 140 EUR
APOSPC440005-250MG
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate
Formel: C₃₆H₁₇F₁₂O₄P
Molecular Weight: 772,49 g/mol
Schmelzpunkt: 162…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689863
CAS-Nummer: 878111-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR