(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-bi-2-naphthol cyclic monophosphate, (11BR)-2,6-bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

PC440006-250MG
APOSPC440006-250MGEA 953 EUR
APOSPC440006-250MG
(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel: C₃₂H₁₇F₄O₄P
Molecular Weight: 572,442 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28139328
CAS-Nummer: 1191451-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR