(S)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-sulphide

Lieferant: Apollo Scientific

PC440008-100MG
APOSPC440008-100MGEA 459 EUR
APOSPC440008-100MG
(S)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-sulphide
(S)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-sulphide
Formel: C₃₆H₁₇F₁₂O₃PS
Molecular Weight: 788,538 g/mol
Schmelzpunkt: 166…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28139330
CAS-Nummer: 1706463-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR